Assay Details
| Gene Symbol: GPR19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003365173.4 | XP_003365221.1 | 461 | 107 | ||
| XM_005610948.3 | XP_005611005.1 | 1131 | 107 | |||
More Information
Set Membership: |
|

GPR19
Genomic Map