Assay Details
| Gene Symbol: TNFAIP8L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001494106.5 | XP_001494156.2 | 2251 | 101 | ||
| XM_023644468.1 | XP_023500236.1 | 1793 | 101 | |||
| XM_023644469.1 | XP_023500237.1 | 1806 | 101 | |||
More Information
Set Membership: |
|

TNFAIP8L1
Genomic Map