Assay Details
| Gene Symbol: CITED2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001496065.6 | XP_001496115.5 | 910 | 99 | ||
| XM_023632890.1 | XP_023488658.1 | 910 | 99 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CITED2| Gene Symbol: CITED2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001496065.6 | XP_001496115.5 | 910 | 99 | ||
| XM_023632890.1 | XP_023488658.1 | 910 | 99 | |||
Set Membership: |
|