Assay Details
| Gene Symbol: LOC102147629 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023650640.1 | XP_023506408.1 | 824 | 104 | ||
| XM_023650641.1 | XP_023506409.1 | 304 | 104 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_023650640.1, XM_023650641.1 transcripts.
Set Membership: |
|

LOC102147629
Genomic Map