Assay Details
| Gene Symbol: PRR15L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005597461.3 | XP_005597518.2 | 481 | 93 | ||
| XM_014740919.2 | XP_014596405.2 | 255 | 93 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PRR15L| Gene Symbol: PRR15L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005597461.3 | XP_005597518.2 | 481 | 93 | ||
| XM_014740919.2 | XP_014596405.2 | 255 | 93 | |||
Set Membership: |
|