Assay Details
| Gene Symbol: ZNF597 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005599017.3 | XP_005599074.1 | 422 | 94 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005599017.3 transcripts.
Set Membership: |
|

ZNF597
Genomic Map