Assay Details
| Gene Symbol: FOXO1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023621529.1 | XP_023477297.1 | 2394 | 106 | ||
| GenBank mRNA | JL638973.1 | - | 1 - 2 | 1042 | 106 | |
More Information
Set Membership: |
|

FOXO1
Genomic Map