Assay Details
| Gene Symbol: DLEU7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001488517.5 | XP_001488567.3 | 1879 | 105 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

DLEU7| Gene Symbol: DLEU7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001488517.5 | XP_001488567.3 | 1879 | 105 | ||
Set Membership: |
|