Assay Details
| Gene Symbol: HOXD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023622044.1 | XP_023477812.1 | 5526 | 68 | ||
| XM_023622045.1 | XP_023477813.1 | 5433 | 68 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

HOXD3| Gene Symbol: HOXD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023622044.1 | XP_023477812.1 | 5526 | 68 | ||
| XM_023622045.1 | XP_023477813.1 | 5433 | 68 | |||
Set Membership: |
|