Assay Details
| Gene Symbol: GPR22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014738918.2 | XP_014594404.1 | 4386 | 103 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_014738918.2 transcripts.
Set Membership: |
|

GPR22
Genomic Map