Assay Details
| Gene Symbol: NMUR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005610708.3 | XP_005610765.1 | 2014 | 59 | ||
| XM_005610709.3 | XP_005610766.1 | 1289 | 59 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

NMUR1| Gene Symbol: NMUR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005610708.3 | XP_005610765.1 | 2014 | 59 | ||
| XM_005610709.3 | XP_005610766.1 | 1289 | 59 | |||
Set Membership: |
|