Assay Details
| Gene Symbol: HIC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023646787.1 | XP_023502555.1 | 149 | 79 | ||
| XM_023646788.1 | XP_023502556.1 | 149 | 79 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_023646787.1, XM_023646788.1 transcripts.
Set Membership: |
|

HIC2
Genomic Map