Assay Details
| Gene Symbol: PWWP2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023636687.1 | XP_023492455.1 | 1939 | 85 | ||
| XM_023636714.1 | XP_023492482.1 | 1835 | 85 | |||
| XM_023636745.1 | XP_023492513.1 | 6209 | 85 | |||
| XM_023636748.1 | XP_023492516.1 | 5630 | 85 | |||
| XM_023636754.1 | XP_023492522.1 | 2085 | 85 | |||
| XM_023636756.1 | XP_023492524.1 | 5450 | 85 | |||
| XM_023636761.1 | XP_023492529.1 | 2036 | 85 | |||
More Information
Set Membership: |
|

PWWP2B
Genomic Map