Assay Details
| Gene Symbol: PINLYP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014727924.2 | XP_014583410.1 | 822 | 99 | ||
| XM_014727967.2 | XP_014583453.1 | 439 | 99 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PINLYP| Gene Symbol: PINLYP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014727924.2 | XP_014583410.1 | 822 | 99 | ||
| XM_014727967.2 | XP_014583453.1 | 439 | 99 | |||
Set Membership: |
|