Assay Details
| Gene Symbol: APOE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005614763.3 | XP_005614820.3 | 219 | 90 | ||
| XM_023649876.1 | XP_023505644.1 | 154 | 90 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: APOE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005614763.3 | XP_005614820.3 | 219 | 90 | ||
| XM_023649876.1 | XP_023505644.1 | 154 | 90 | |||
Set Membership: |
|