Assay Details
| Gene Symbol: SNRPD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001502749.5 | XP_001502799.1 | 123 | 72 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001502749.5 transcripts.
Set Membership: |
|

SNRPD2
Genomic Map