Assay Details
| Gene Symbol: PPP1R15A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005596389.3 | XP_005596446.2 | 245 | 125 | ||
| XM_023650066.1 | XP_023505834.1 | 245 | 125 | |||
| XM_023650067.1 | XP_023505835.1 | 245 | 125 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005596389.3, XM_023650066.1, XM_023650067.1 transcripts.
Set Membership: |
|

PPP1R15A
Genomic Map