Assay Details
| Gene Symbol: NDUFA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001488297.6 | XP_001488347.2 | 316 | 59 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001488297.6 transcripts.
Set Membership: |
|

NDUFA3
Genomic Map