Assay Details
| Gene Symbol: LOC100054684 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005596530.3 | XP_005596587.2 | 1642 | 80 | ||
| XM_014730730.2 | XP_014586216.2 | 1613 | 80 | |||
More Information
Set Membership: |
|

LOC100054684
Genomic Map