Assay Details
| Gene Symbol: ZNF550 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014731213.2 | XP_014586699.2 | 2735 | 96 | ||
| XM_023650589.1 | XP_023506357.1 | 2644 | 96 | |||
More Information
Set Membership: |
|

ZNF550
Genomic Map