Assay Details
| Gene Symbol: ATP5PD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001496541.4 | XP_001496591.1 | 284 | 106 | ||
| XM_005597174.3 | XP_005597231.1 | 266 | 106 | |||
| XM_005597175.3 | XP_005597232.1 | 262 | 106 | |||
| XM_005597176.3 | XP_005597233.1 | 218 | 106 | |||
| XM_023652058.1 | XP_023507826.1 | 203 | 106 | |||
| GenBank mRNA | JL619249.1 | - | 1 - 2 | 134 | 105 | |
More Information
Set Membership: |
|

ATP5PD
Genomic Map