Assay Details
| Gene Symbol: GPR142 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014736594.2 | XP_014592080.2 | 2338 | 86 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

GPR142| Gene Symbol: GPR142 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014736594.2 | XP_014592080.2 | 2338 | 86 | ||