Assay Details
| Gene Symbol: LOC100063851 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005597306.3 | XP_005597363.3 | 5976 | 59 | ||
| XM_005597307.3 | XP_005597364.3 | 5904 | 59 | |||
More Information
Set Membership: |
|

LOC100063851
Genomic Map