Assay Details
| Gene Symbol: LOC100063851 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005597306.3 | XP_005597363.3 | 3462 | 158 | ||
| XM_005597307.3 | XP_005597364.3 | 3462 | 158 | |||
More Information
Set Membership: |
|

LOC100063851
Genomic Map