Assay Details
| Gene Symbol: LOC100073135 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023653539.1 | XP_023509307.1 | 286 | 98 | ||
| XM_023653540.1 | XP_023509308.1 | 240 | 98 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100073135| Gene Symbol: LOC100073135 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023653539.1 | XP_023509307.1 | 286 | 98 | ||
| XM_023653540.1 | XP_023509308.1 | 240 | 98 | |||