Assay Details
| Gene Symbol: RBAK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023655209.1 | XP_023510977.1 | 743 | 106 | ||
| XM_023655210.1 | XP_023510978.1 | 434 | 106 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: RBAK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023655209.1 | XP_023510977.1 | 743 | 106 | ||
| XM_023655210.1 | XP_023510978.1 | 434 | 106 | |||
Set Membership: |
|