Assay Details
| Gene Symbol: TRIM4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023616801.1 | XP_023472569.1 | 1172 | 77 | ||
| XM_023616802.1 | XP_023472570.1 | 1252 | 77 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TRIM4| Gene Symbol: TRIM4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023616801.1 | XP_023472569.1 | 1172 | 77 | ||
| XM_023616802.1 | XP_023472570.1 | 1252 | 77 | |||
Set Membership: |
|