Assay Details
Gene Symbol: NYAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005598626.3 | XP_005598683.2 | 1736 | 90 | ||
XM_023655343.1 | XP_023511111.1 | 609 | 90 | |||
XM_023655344.1 | XP_023511112.1 | 1199 | 90 | |||
XM_023655345.1 | XP_023511113.1 | 2851 | 90 | |||
XM_023655346.1 | XP_023511114.1 | 2222 | 90 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
