Assay Details
| Gene Symbol: ZNF213 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005599037.3 | XP_005599094.2 | 969 | 71 | ||
| XM_014730208.2 | XP_014585694.2 | 1351 | 71 | |||
| XM_023616463.1 | XP_023472231.1 | 1765 | 71 | |||
| XM_023616464.1 | XP_023472232.1 | 3186 | 71 | |||
More Information
Set Membership: |
|

ZNF213
Genomic Map