Assay Details
| Gene Symbol: LOC100630872 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014730426.2 | XP_014585912.1 | 160 | 157 | ||
| XM_014730427.2 | XP_014585913.1 | 164 | 82 | |||
More Information
Set Membership: |
|

LOC100630872
Genomic Map