Assay Details
| Gene Symbol: LOC106781592 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023617474.1 | XP_023473242.1 | 345 | 85 | ||
| XM_023617475.1 | XP_023473243.1 | 341 | 85 | |||
More Information
Set Membership: |
|

LOC106781592
Genomic Map