Assay Details
| Gene Symbol: LOC100072343 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023617533.1 | XP_023473301.1 | 10560 | 74 | ||
| XM_023617534.1 | XP_023473302.1 | 6322 | 74 | |||
| XM_023617535.1 | XP_023473303.1 | 5108 | 74 | |||
| XM_023617536.1 | XP_023473304.1 | 7530 | 74 | |||
| XM_023617537.1 | XP_023473305.1 | 5293 | 74 | |||
| XM_023617538.1 | XP_023473306.1 | 10563 | 74 | |||
| XM_023617539.1 | XP_023473307.1 | 5520 | 74 | |||
| XM_023617540.1 | XP_023473308.1 | 3872 | 74 | |||
| XM_023617541.1 | XP_023473309.1 | 7291 | 74 | |||
| XM_023617542.1 | XP_023473310.1 | 5733 | 74 | |||
| XM_023617543.1 | XP_023473311.1 | 4135 | 74 | |||
| XM_023617544.1 | XP_023473312.1 | 7100 | 74 | |||
More Information
Set Membership: |
|

LOC100072343
Genomic Map