Assay Details
| Gene Symbol: TUBB6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023648220.1 | XP_023503988.1 | 374 | 77 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TUBB6| Gene Symbol: TUBB6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023648220.1 | XP_023503988.1 | 374 | 77 | ||
Set Membership: |
|