Assay Details
| Gene Symbol: GPR173 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014728929.2 | XP_014584415.2 | 3518 | 71 | ||
| XM_023633366.1 | XP_023489134.1 | 3661 | 71 | |||
More Information
Set Membership: |
|

GPR173
Genomic Map