Assay Details
| Gene Symbol: LEAP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003362819.4 | XP_003362867.1 | 4115 | 96 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LEAP2| Gene Symbol: LEAP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003362819.4 | XP_003362867.1 | 4115 | 96 | ||