Assay Details
| Gene Symbol: OCM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014729796.2 | XP_014585282.1 | 1130 | 89 | ||
| XM_023655261.1 | XP_023511029.1 | 605 | 89 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: OCM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014729796.2 | XP_014585282.1 | 1130 | 89 | ||
| XM_023655261.1 | XP_023511029.1 | 605 | 89 | |||
Set Membership: |
|