Assay Details
| Gene Symbol: TMEM235 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014736488.2 | XP_014591974.1 | 931 | 74 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TMEM235| Gene Symbol: TMEM235 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014736488.2 | XP_014591974.1 | 931 | 74 | ||