Assay Details
| Gene Symbol: LOC100049849 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001487836.3 | XP_001487886.1 | 958 | 107 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100049849| Gene Symbol: LOC100049849 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001487836.3 | XP_001487886.1 | 958 | 107 | ||
Set Membership: |
|