Assay Details
| Gene Symbol: GCNT4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023618168.1 | XP_023473936.1 | 121 | 187 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_023618168.1 transcripts.
Set Membership: |
|

GCNT4
Genomic Map