Assay Details
| Gene Symbol: CHST13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023619843.1 | XP_023475611.1 | 827 | 75 | ||
| XM_023619844.1 | XP_023475612.1 | 201 | 75 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CHST13| Gene Symbol: CHST13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023619843.1 | XP_023475611.1 | 827 | 75 | ||
| XM_023619844.1 | XP_023475612.1 | 201 | 75 | |||
Set Membership: |
|