Assay Details
| Gene Symbol: DNAJC22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001492026.5 | XP_001492076.4 | 1124 | 92 | ||
| XM_014741015.2 | XP_014596501.1 | 1199 | 92 | |||
| XM_023643488.1 | XP_023499256.1 | 1245 | 92 | |||
| XM_023643489.1 | XP_023499257.1 | 2316 | 92 | |||
| XM_023643490.1 | XP_023499258.1 | 1245 | 92 | |||
| XM_023643491.1 | XP_023499259.1 | 1123 | 92 | |||
More Information
Set Membership: |
|

DNAJC22
Genomic Map