Assay Details
| Gene Symbol: DYNLT4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023631248.1 | XP_023487016.1 | 326 | 77 | ||
| XM_023631257.1 | XP_023487025.1 | 317 | 77 | |||
| XM_023631277.1 | XP_023487045.1 | 203 | 77 | |||
| XM_023631295.1 | XP_023487063.1 | 164 | 77 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_023631277.1, XM_023631295.1 transcripts.
Set Membership: |
|

DYNLT4
Genomic Map