Assay Details
| Gene Symbol: GJE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023632999.1 | XP_023488767.1 | 160 | 94 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_023632999.1 transcripts.
Set Membership: |
|

GJE1
Genomic Map