Assay Details
| Gene Symbol: RP1L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023636250.1 | XP_023492018.1 | 91 | 74 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_023636250.1 transcripts.
Set Membership: |
|

RP1L1
Genomic Map