Assay Details
| Gene Symbol: CCDC71L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023638880.1 | XP_023494648.1 | 814 | 95 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_023638880.1 transcripts.
Set Membership: |
|

CCDC71L
Genomic Map