Assay Details
| Gene Symbol: PTGER1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023645208.1 | XP_023500976.1 | 1119 | 72 | ||
| XM_023645209.1 | XP_023500977.1 | 1082 | 72 | |||
More Information
Set Membership: |
|

PTGER1
Genomic Map