Assay Details
| Gene Symbol: LYL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023645289.1 | XP_023501057.1 | 583 | 84 | ||
| XM_023645290.1 | XP_023501058.1 | 1535 | 84 | |||
| XM_023645291.1 | XP_023501059.1 | 647 | 84 | |||
More Information
Set Membership: |
|

LYL1
Genomic Map