Assay Details
| Gene Symbol: NKPD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023649882.1 | XP_023505650.1 | 2899 | 54 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

NKPD1| Gene Symbol: NKPD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023649882.1 | XP_023505650.1 | 2899 | 54 | ||
Set Membership: |
|