Assay Details
| Gene Symbol: LOC102147628 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023634374.1 | XP_023490142.1 | 521 | 100 | ||
| XM_023634375.1 | XP_023490143.1 | 518 | 100 | |||
More Information
Set Membership: |
|

LOC102147628
Genomic Map