Assay Details
| Gene Symbol: NUDT15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004938701.3 | XP_004938758.1 | 1 - 2 | 1847 | 85 | |
| XM_417053.6 | XP_417053.4 | 1 - 2 | 1847 | 85 | ||
| GenBank mRNA | BX929709.1 | - | 1 - 2 | 201 | 85 | |
| CR386374.1 | - | 1 - 2 | 208 | 85 | ||
More Information
Set Membership: |
|

NUDT15
Genomic Map